• USA Home
  • 20259 - ORBO 33 Activated Petroleum Charcoal (20/40), 700/390 mg

EMAIL THIS PAGE TO A FRIEND
20259 Supelco

ORBO 33 Activated Petroleum Charcoal (20/40), 700/390 mg

W,F,F separators, O.D. × L 8 mm × 150 mm, pkg of 50 ea

Synonym: Petroleum Charcoal Tube

Purchase

Properties

Related Categories Active Sampling, Air Monitoring, Analytical/Chromatography, Charcoal Sorbent Tubes, ORBO Sorbent Tubes For Gas & Vapor Sampling,
material   W,F,F separators
composition   Bed A, 700 mg
  Bed B, 390 mg
mfr. no.   ORBO 33
packaging   pkg of 50 ea
O.D. × L   8 mm × 150 mm
matrix   Activated petroleum charcoal
  petroleum charcoal charcoal
particle size   20-40 mesh
  20-40 mesh
compatibility   mode of use Solvent desorption
  technique used for Air monitoring

Description

Legal Information

ORBO is a trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water

Non-Sterile Millex Syringe Filters
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?