• USA Home
  • 20264-U - ORBO 42 Large Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 100/50 mg

EMAIL THIS PAGE TO A FRIEND
20264-U Supelco

ORBO 42 Large Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 100/50 mg

W,W,W (specially treated) separators, O.D. × L 10 mm × 100 mm, pkg of 50 ea

Synonym: ORBO Supelpak 20 series Tube

Purchase

Properties

Related Categories Active Sampling, Air Monitoring, Analytical/Chromatography, ORBO Sorbent Tubes For Gas & Vapor Sampling, Porous Polymer Sorbent Tubes,
material   W,W,W (specially treated) separators
composition   Bed A, 100 mg
  Bed B, 50 mg
mfr. no.   ORBO 42 Large
packaging   pkg of 50 ea
O.D. × L   10 mm × 100 mm
matrix   XAD-2 (specially treated)
particle size   20-40 mesh
suitability   suitable for 2550 per NIOSH

Description

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

ORBO is a trademark of Sigma-Aldrich Co. LLC

Supelpak is a trademark of Sigma-Aldrich Co. LLC

XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?