• USA Home
  • 20274 - Carbotrap®/Carbopack Adsorbent

EMAIL THIS PAGE TO A FRIEND
20274 Supelco

Carbotrap®/Carbopack Adsorbent

60-80 mesh, matrix Carbopack B HT, bottle of 10 g

Purchase

Properties

packaging   bottle of 10 g
matrix   Carbopack B HT
particle size   60-80 mesh
pore size   ~0 cm3/g macroporosity
  ~0 cm3/g mesoporosity
  ~0 cm3/g microporosity
  ~0 Å pore diameter
surface area   ~100 m2/g
density   ~0.36 g/mL (free fall density)

Description

Legal Information

Carbopack is a trademark of Sigma-Aldrich Co. LLC

Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
FF5800000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?