EMAIL THIS PAGE TO A FRIEND
20438-U Supelco

Plastic Column Cap

for use with 3/16 in. O.D. column

Purchase

Properties

Related Categories Analytical/Chromatography, Column Caps, GC Accessories, Gas Chromatography (GC), Packed Column Preparation, Installation, Maintenance, and Storage More...
packaging   pkg of 100 ea
fitting   cap female slip-on for column 3/16 in. O.D.
compatibility   for use with 3/16 in. O.D. column

Description

General description

These plastic caps will easily slip over the ends of packed GC columns to protect them during storage.

Application

Plastic column cup maybe used to close the immunoaffinity chromatography column used for separating cross-linked protein complexes adhering to P1 protein of Mycoplasma pneumonia.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?