• USA Home
  • 20528-U - Tubing SS 3/16 in. 0D x 3.7 mm ID 50ft coil
EMAIL THIS PAGE TO A FRIEND
20528-U Supelco
Tubing SS 3/16 in. 0D x 3.7 mm ID 50ft coil
tubing L × O.D. × I.D. 50 ft × 3/16 in. × 3.7 mm
Purchase
Properties
Related Categories Analytical/Chromatography, Gas Chromatography (GC), Gas Purification and Gas Management, Plumbing/Regulation, Premium Grade 304 Stainless Steel,
material   regular grade stainless steel (304)
tubing L × O.D. × I.D.   50 ft × 3/16 in. × 3.7 mm
Description
Application
304SS tubing may be used to make homemade reactor coil for Gas chromatography oven to determine the effect of thermal volume expansion of solvents under superheated conditions.
General description
Type 304 stainless steel is covered by highly protective film of chromium oxyhydroxide. Hydrochloric acid solution readily attacks 304SS. Hence, it is recommended to add inhibitors like heterocyclic compounds to reduce the corrosion rate. 304SS materials are suitable for tubing and for wires as they exhibit strength and are corrosion resistant.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?