• USA Home
  • 20631 - Oxisorb® Oxygen Scrubber

EMAIL THIS PAGE TO A FRIEND
20631 Supelco

Oxisorb® Oxygen Scrubber

Replacement cartridge

  •  eCl@ss 32110502

Purchase

Description

General description

This trap will reduce oxygen and moisture to less than 1 ppm when the incoming level of oxygen is below 15 ppm and the incoming level of moisture is below 10 ppm. Use alternative traps when the levels in the incoming gas exceed these values. The kit includes a cartridge, and fittings for installation. Replacing the cartridge is simple, and can be performed quickly.

Dimensions:
• Cartridge and Fittings: 91/4 in. (23.5 cm) long x 2 in. (51 mm) diameter
• Cartridge Only: 47/8 in. (12.4 cm) long x 11/8 in. (29 mm) diameter

Application

N2, CO2 and CO2-N2 gas mixture may be passed through Oxisorb® Oxygen Scrubber before being used in an experimental study, done to monitor the kinetics of sulphate and acetate uptake, by the sulphate-reducing bacteria Desulfobacter postgatei.

Legal Information

Oxisorb is a registered trademark of Messer Group GmbH

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
GB6650000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?