• USA Home
  • 20644-U - Supeltex® M-2 Packed Column Ferrule, 1/16 in. Column O.D.

EMAIL THIS PAGE TO A FRIEND
20644-U Supelco

Supeltex® M-2 Packed Column Ferrule, 1/16 in. Column O.D.

I.D. 1/16 in., 1/16 in. O.D. Column, pkg of 10 ea

Purchase

Properties

Related Categories Analytical/Chromatography, Column Ferrules, Nuts, and Specialized Hand Tools, GC Accessories, Gas Chromatography (GC), Packed GC Ferrules, 1/16 in. O.D. Columns,
packaging   pkg of 10 ea
maximum temperature   350 °C
I.D.   1/16 in.
compatibility   1/16 in. O.D. Column

Description

General description

Characteristics: High reusability.

Material Composition: DuPont VESPEL SP-1 (100% polyimide)

Maximum Temperature:
350 °C

Legal Information

Supeltex is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?