EMAIL THIS PAGE TO A FRIEND
20881 Supelco

Carbotrap® 317

glass TD tube (fast-flow), preconditioned

Purchase

Properties

Related Categories Active Sampling, Air Monitoring, Analytical/Chromatography, Dynatherm Fast Flow Sampling Tubes, Sample Preparation & Purification,
material   glass TD tube (fast-flow)
feature   preconditioned
packaging   pkg of 1 ea
I.D. × O.D. × L   7 mm × 10 mm × 4.5 in.
matrix   Carbotrap® 317
suitability   suitable for TO-17 per US EPA
compatibility   for use with Dynatherm ACEM-9300, IACEM-980, ACEM 900 and MTDU equipped with fast-flow "FF"

Description

Application

TO-17 compounds and other volatile and semivolatile compounds in air

Legal Information

Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC

Dynatherm is a trademark of CDS Analytical

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles

Articles

Thermal Desorption Tube Selection Guide

In addition to the general guidelines presented in the tables below view the technical report titled “A Tool for Selecting an Adsorbent for Thermal Desorption Applications” (1,770Kb pdf - T402025), w...
Keywords: Sample preparations

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?