• USA Home
  • 20949 - Precision Sampling syringe needles
EMAIL THIS PAGE TO A FRIEND
20949 Supelco
Precision Sampling syringe needles
volume size 5‑10 μL, for use with VICI Series CG-130, stainless steel needle, needle L × O.D. × I.D. 2.25 in. × 0.019 in. × 0.005 in.
Synonym: XTRA NEEDLES C120,C160,CG130 PK3
Purchase
Properties
Related Categories Analytical/Chromatography, Syringes, VICI (Valco) Precision Sampling Syringes, VICI Replacement Needles
material   stainless steel needle
volume size   5‑10 μL
description   CG-130
feature   bevel tip (Point style 2)
packaging   pkg of 3 ea
mfr. no.   VICI, 123050
needle L × O.D. × I.D.   2.25 in. × 0.019 in. × 0.005 in.
compatibility   for use with VICI Series CG-130
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?