• USA Home
  • 20955-U - Valco® Precision Sampling syringe, series CG-130

EMAIL THIS PAGE TO A FRIEND
20955-U Supelco

Valco® Precision Sampling syringe, series CG-130

volume 10 μL, CG-130, stainless steel needle bevel tip (point style 2), needle type, Fixed, needle L 2 in.

Synonym: SYRINGE,PS-CG-130-FN,10U

Purchase

Properties

Related Categories Analytical/Chromatography, Liquid syringes, Series CG-130, Syringes, VICI (Valco) Precision Sampling Syringes More...
material   stainless steel needle bevel tip (point style 2)
needle size   (bevel point)
feature   needle type Fixed
  syringe type CG-130
packaging   pkg of 1 ea
mfr. no.   VICI, 130022
needle L   2 in.
volume   10 μL

Description

General description

CG-130: This model has a reinforced plunger syringe with an internal guide and special plunger that eliminates bending
problems.

Legal Information

Valco is a registered trademark of Valco Instruments Co. Inc.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?