• USA Home
  • 21776 - Empty Glass GC Column, for Agilent® 5880, 5890, 6890, 7820, and 7890 (Configuration "A")
EMAIL THIS PAGE TO A FRIEND
21776 Supelco
Empty Glass GC Column, for Agilent® 5880, 5890, 6890, 7820, and 7890 (Configuration "A")
L × O.D. × I.D. 4.0 ft (1.2 m) × 1/4 in. × 2.0 mm
Purchase
Properties
Related Categories Analytical/Chromatography, Empty GC Columns, Empty Glass GC Columns, Empty Glass GC Columns, for Agilent 5880, 5890, 6890, 7820, and 7890 (Configuration A), Gas Chromatography (GC),
material   glass column
application(s)   gas chromatography (GC): suitable
L × O.D. × I.D.   4.0 ft (1.2 m) × 1/4 in. × 2.0 mm
column type   empty
Description
General description
Silane treated empty glass column. Fittings are not included.
• X (length of injector arm) = 11.02 in. (280 mm)
• Y (length of detector arm) = 9.05 in. (230 mm)
• S (span, injector to detector) = 9.0 in. (229 mm)
This column fits Agilent 5880, 5890, 6890, 7820, and 7890 (configuration A, on-column injection, all detectors except TCD).
GC columns are of two types; packed column and capillary column. Capillary column carries the gas through a central aperture which is unimpeded through the entire length of column. In general, capillary columns are made longer to produce more theoretical plates. This also helps in band dispersion eventually narrowing the peaks and resulting in high resolution.
Application
Empty column may be used for packing adsorbents for temperature-programmed desorption (TPD) instrumentation.
Legal Information
Agilent is a registered trademark of Agilent Technologies, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?