EMAIL THIS PAGE TO A FRIEND
22051-U Supelco

Swagelok® Union

Swagelok®, 100-6, stainless steel, 1/16 in.

Synonym: Swagelok® 100-6,SS UNION 1/16" EA

  •  eCl@ss 32035302

Purchase

Properties

Related Categories Analytical/Chromatography, Gas Chromatography (GC), Gas Purification and Gas Management, Plumbing/Regulation, Swagelok Tubing Fitting,
material   stainless steel
packaging   pkg of 1 ea
mfr. no.   Swagelok®, 100-6
fitting   1/16 in.

Description

General description

Nut and ferrules included.

Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?