• USA Home
  • 22100-U - Swagelok® Pipe Adapter

EMAIL THIS PAGE TO A FRIEND
22100-U Supelco

Swagelok® Pipe Adapter

Swagelok®, 4-TA-1-2, brass, 1/4 in. tube, 1/8 in. male NPT

Synonym: Swagelok® 4-TA-1-2,BRASS PIPE ADAPT 1/4X1/8 PK2

Purchase

Properties

Related Categories Analytical/Chromatography, Gas Chromatography (GC), Gas Purification and Gas Management, Plumbing/Regulation, Swagelok Pipe Adapter,
material   brass
packaging   pkg of 2 ea
mfr. no.   Swagelok®, 4-TA-1-2
fitting   1/4 in. tube
  1/8 in. male NPT

Description

Legal Information

Swagelok is a registered trademark of Swagelok Company

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?