• USA Home
  • 22125-U - On/Off Toggle Valve

EMAIL THIS PAGE TO A FRIEND
22125-U Supelco

On/Off Toggle Valve

brass, angle arms 1/4 in.

Synonym: Swagelok® 1GS4-A,BRASS TOGGLE VALVE 1/4X1/4A EA

  •  eCl@ss 32035302

Purchase

Description

General description

Nuts and ferrules included.

Toggle valve is one of the most simple and widely used industry valve. It′s on and off settings determines whether or not fluid flows past the valve. There are many styles of toggle valve like ball valve, gate valve, plug valve and knife valve.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?