• USA Home
  • 22140-U - On/Off Throttling Valve

EMAIL THIS PAGE TO A FRIEND
22140-U Supelco

On/Off Throttling Valve

brass, 1/4 in.

Synonym: Swagelok® 4P4T,BRASS THROTTLING VALVE 1/4" EA

  •  eCl@ss 32035302

Purchase

Description

General description

Nuts and ferrules included.

Throttling valve′s on and off settings determines whether or not fluid flows past the valve. It is much more efficient than the toggle on/off valve, as it provides better flow control between 0 and 100% flow and has a larger baseline pressure drop. There are many styles of throttling valve like globe valve and butterfly valve.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?