• USA Home
  • 22465 - Supeltex® M-2 Double-Tapered Ferrule

EMAIL THIS PAGE TO A FRIEND
22465 Supelco

Supeltex® M-2 Double-Tapered Ferrule

I.D. 0.4-0.8 mm, 0.10-0.25 mm I.D. to 0.53 mm I.D. column (reducing), pkg of 2 ea

Purchase

Properties

Related Categories Analytical/Chromatography, Capillary Column Butt Connectors, Capillary Column Connectors, Capillary GC Columns and Guard Columns/Retention Gaps, GC Accessories,
feature   ferrule type double-tapered
packaging   pkg of 2 ea
application(s)   gas chromatography (GC): suitable
I.D.   0.4-0.8 mm
compatibility   0.10-0.25 mm I.D. to 0.53 mm I.D. column (reducing)

Description

General description

• Max. Temp.: 350 °C
• Composition: DuPont VESPEL SP-1 (100% polyimide)
• Characteristics: High reusability.

Legal Information

Supeltex is a registered trademark of Sigma-Aldrich Co. LLC

Other Notes

Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

How to Choose a Capillary GC Column

An optimized chromatographic separation begins with the column. The selection of the proper capillary column for any application should be based on four significant factors: stationary phase, column ...
How to Choose a Capillary GC Column
Keywords: Chromatography, Condensations, Gas chromatography, Immobilization, Mass spectrometry, Pesticides, Separation, Size-exclusion chromatography, Solid phase microextractions, Solvents, Vaporization

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?