EMAIL THIS PAGE TO A FRIEND
22662 Supelco

Immersion Cleaner

Purchase

Properties

packaging   bottle of 100 mL
concentration   10 × in H2O (mix 1:10 for use)

Description

General description

For dirty detectors.
An aqueous and nontoxic surfactant solution that removes heavy deposits of silica from a detector.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

22404 Packed GC FID Cleaning Kit, for use with Varian instruments
22403 Packed GC FID Cleaning Kit, for use with Agilent/HP instruments

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?