• USA Home
  • 22684 - Swagelok® Union Cross

EMAIL THIS PAGE TO A FRIEND
22684 Supelco

Swagelok® Union Cross

Swagelok®, 200-4, brass, 1/8 in. Swagelok

Synonym: Swagelok® 200-4,BRASS UNION CROSS 1/8" EA

  •  eCl@ss 32035302

Purchase

Properties

Related Categories Analytical/Chromatography, Gas Chromatography (GC), Gas Purification and Gas Management, Plumbing/Regulation, Swagelok Tubing Fitting,
material   brass
packaging   pkg of 1 ea
mfr. no.   Swagelok®, 200-4
fitting   1/8 in. Swagelok

Description

General description

Nuts and ferrules included.

Union cross generally divides the eluent flow into three directions which is coming from the pump. In gas system, pure gases from mass flow controllers are converged at the union cross and in turn flow into different size tubing to generate turbulence which produce homogenized gas.

Application

Swagelok® union cross was used in sensor test module along with electrical breakout box to pressurized high temperature reactors such as high performance reactor (HPR) microreactor.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?