23139 Supelco
Magnifier
Coddington Magnifier
DOWNLOAD MSDS (PDF)
Properties
Related Categories Analytical/Chromatography, Hand Tools, Illuminated Magnifiers, Laboratory Equipment, Laboratory Supplies More...
feature   lens 20 X (single thick lens)
packaging   pkg of 1 ea
Description
General description
Reading glass is 75 mm diam. with metal handle. Folding style has 3 × 25 mm glass lenses in a plastic case.
Second only to the Hastings triplet in image quality. The 20 × image is provided by a single thick lens (rather than a three-component lens). Swing-away nickel-plated case. No built-in illumination source.
Price and Availability
Customers Also Viewed
Magnifier
Z500674Magnifier
Folding
Magnifier
Z500569Magnifier
Reading
Magnifier
Lenscope Illuminated Magnifier
Magnifier
Compact Lighted Magnifier
Scale for Lenscope Magnifier, 360° in 1° intervals
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?