• USA Home
  • 24230-U - SPB®-1 Capillary GC Column

EMAIL THIS PAGE TO A FRIEND
24230-U Supelco

SPB®-1 Capillary GC Column

L × I.D. 25 m × 0.20 mm, df 0.33 μm

  •  eCl@ss 32119290

Purchase

Properties

Related Categories Analytical/Chromatography, Capillary GC Columns and Guard Columns/Retention Gaps, Capillary GC Columns, by name, Gas Chromatography (GC), SPB-1 More...
material   fused silica
Beta value   152
df   0.33 μm
application(s)   gas chromatography (GC): suitable
L × I.D.   25 m × 0.20 mm
matrix active group   Bonded; poly(dimethyl siloxane) phase
column type   capillary non-polar

Description

General description

Application: This column is often used for traditional general purpose applications, where a non-polar column is required. Analytes will be separated primarily according to boiling point.
USP Code: This column meets USP G1, G2, and G9 requirements.
Phase:
• Bonded
• Poly(dimethyl siloxane)
Temp. Limits:
• ≤0.32 mm I.D., <2 μm: -60 °C to 320 °C (isothermal or programmed)
• ≤0.32 mm I.D., ≥2 μm: -60 °C to 300 °C (isothermal or programmed)
• ≥0.53 mm I.D., <2 μm: -60 °C to 300 °C (isothermal) or 320 °C (programmed)
• ≥0.53 mm I.D., ≥2 μm: -60 °C to 260 °C (isothermal) or 280 °C (programmed)

Application

PAG Capillary GC Column may be used for the separation of aliphatic alcohols on a polar stationary phase.

Legal Information

SPB is a registered trademark of Sigma-Aldrich Co. LLC

Other Notes

Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Related Content

How to Choose a Capillary GC Column

An optimized chromatographic separation begins with the column. The selection of the proper capillary column for any application should be based on four significant factors: stationary phase, column ...
How to Choose a Capillary GC Column
Keywords: Chromatography, Condensations, Gas chromatography, Immobilization, Mass spectrometry, Pesticides, Separation, Size-exclusion chromatography, Solid phase microextractions, Solvents, Vaporization

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?