• USA Home
  • 24406 - SGE Syringes for PerkinElmer® Autosystem
EMAIL THIS PAGE TO A FRIEND
24406 Supelco
SGE Syringes for PerkinElmer® Autosystem
volume 0.5 μL, 0.5BR-PE-0.47, removable needle, needle size 26 ga (cone tip), needle L 70 mm
Purchase
Properties
Related Categories Analytical/Chromatography, GC Autosampler Syringes, PerkinElmer AS100, AS200, AS3000 GC Autosystems, Syringes
material   stainless steel needle cone tip (point style 1)
needle size   26 ga (cone tip)
description   color (light orange)
feature   removable needle
packaging   pkg of 1 ea
mfr. no.   SGE, 000475
needle L   70 mm
volume   0.5 μL
Description
Legal Information
PerkinElmer is a registered trademark of PerkinElmer Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?