• USA Home
  • 24552 - Containers, precleaned
EMAIL THIS PAGE TO A FRIEND
24552 Supelco
Containers, precleaned
volume 236 mL, clear glass vial, tall, wide mouth, thread 58-400, pkg of 24 ea
Purchase
Properties
Related Categories Analytical/Chromatography, Precleaned Containers, Silanized & Precleaned Vials, Vials
material   clear glass vial
feature   closure type black polypropylene solid cap, PTFE liner
  precleaned
  tall, wide mouth
packaging   pkg of 24 ea
volume   236 mL
weight   227 g (container)
fitting   thread 58-400
Description
Frequently Asked Questions
Frequently Asked Questions are available for this Product.
General description
The following containers are cleaned according to US EPA protocol B. Black polypropylene cap with 0.030" PTFE liner.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Frequently Asked Questions
Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis. These documents are located on the product detail page under Useful Links & Tools. Click on the following link to search for a Certificate of Analysis. Please click the following link to see the details on our Product Dating Information.
How do I get lot-specific information or a Certificate of Analysis?
A Certificate of Analysis is available by lot number and can be obtained through our Advanced Search Option: http://www.sigmaaldrich.com/catalog/AdvancedSearchPage.do
What steps are used to prepare Precleaned containers, Product 24552, according to US EPA Protocol B?
Wash containers in hot water using laboratory grade non-phosphate detergent Rinse three times with copious amounts of tap water to remove detergent Rinse three times with ASTM Type I organic-free water Oven-dry containers Allow containers to cool to room temperature in an enclosed contaminant-free environment
Are organic solvents used in the US EPA Protocol B cleaning procedure?
No organic solvents are used in the cleaning process.
Is a Certificate of Analysis available for Product No. 24552 Containers, Precleaned, size 8oz?
Product No. 24552 is shipped with a Certificate of Analysis.
How do I find price and availability?
There are several ways to find pricing and availability for our products.  Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers. 
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS. To access the shipping information for this material, use the link on the product detail page for the product, or search here. 
My question is not addressed here, how can I contact Technical Service for assistance?
Use the option to the right to "Ask a Question" by email of a Technical Service Scientist.
Show more questions
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
24551-U Containers, precleaned, volume 118 mL, clear glass vial, tall, wide mouth, thread 48-400, pkg of 24 ea
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?