• USA Home
  • 24760-U - Closures for Snap Ring vials
EMAIL THIS PAGE TO A FRIEND
24760-U Supelco
Closures for Snap Ring vials
polypropylene seal, PTFE/silicone/PTFE, for 11 mm snap ring, pkg of 100 ea
Purchase
Properties
Related Categories Analytical/Chromatography, Autosampler Vials, Caps, Crimp Seals and Stoppers, Poly-Crimp Seals for 2 mL vials, Snap Caps,
material   PTFE/silicone/PTFE
  polypropylene seal
feature   closure type snap ring seal
packaging   pkg of 100 ea
O.D. × H × I.D.   11.5 mm × 6 mm × 9 mm
fitting   for 11 mm snap ring
compatibility   for use with Large Opening Crimp/Snap Seal Vials
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?