• USA Home
  • 24951 - Fused Silica-Lined Stainless Steel Tubing
EMAIL THIS PAGE TO A FRIEND
24951 Supelco
Fused Silica-Lined Stainless Steel Tubing
tubing L × O.D. × I.D. 25 ft × 1/16 in. (1.59 mm) × 0.01 in. (0.254 mm)
  •  eCl@ss 32119290
Purchase
Properties
Related Categories Analytical/Chromatography, Fused Silica-Lineed Stainless Steel Tubing, Gas Chromatography (GC), Gas Purification and Gas Management, Plumbing/Regulation,
tubing L × O.D. × I.D.   25 ft × 1/16 in. (1.59 mm) × 0.01 in. (0.254 mm)
Description
General description
The strength of stainless steel with the inertness of deactivated fused silica. Use for gas transfer lines.
Application
Fused silica-lined stainless steel tubing may be used in the measurement of volatile organic compounds (VOCs) during air quality study using VOC sampling system.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?