EMAIL THIS PAGE TO A FRIEND
25355 Supelco

PTE-5

L × I.D. 15 m × 0.53 mm, df 0.50 μm

Purchase

Properties

material   fused silica
description   GC capillary column
Beta value   265
df   0.50 μm
application(s)   gas chromatography (GC): suitable
L × I.D.   15 m × 0.53 mm
matrix active group   bonded; poly(5% diphenyl/95% dimethylsiloxane) phase
McReynolds constant   19, 74, 64, 93, 62

Description

Chem/Phys Resistance

Temp. Limits:
• -60 °C to 320 °C

General description

PTE-5/QTM - is a special purpose low bleed column for use with GC-MS systems and was designed for use with US EPA Methods 625, 1625, 8270 semivolatile pollutants, and QTM protocols. Because the Quick Turnaround Method (QTM) version of the PTE-5 column length is only 15 meters, this column is often used for fast analyses.

A special purpose low bleed column for use with GC-MS systems and was designed for use with US EPA Methods 625, 1625, 8270 semivolatile pollutants, and QTM protocols. Because the Quick Turnaround Method (QTM) version of the PTE-5 column length is only 15 meters, this column is often used for fast analyses.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?