• USA Home
  • 26003 - Glass Packed GC Column (fits Agilent, Configuration "A")

EMAIL THIS PAGE TO A FRIEND
26003 Supelco

Glass Packed GC Column (fits Agilent, Configuration "A")

phase 0.1% SP-1000, matrix 80/100 Carbopack C support, L × O.D. × I.D. 6.6 ft (2.0 m) × 1/4 in. × 2.0 mm, TightSpec

Purchase

Properties

Related Categories Analytical/Chromatography, Gas Chromatography (GC), Glass Packed GC Columns (fit Agilent, Configuration "A"), Packed GC Columns, Packed GC Columns and Components More...
material   glass column
application(s)   gas chromatography (GC): suitable
L × O.D. × I.D.   6.6 ft (2.0 m) × 1/4 in. × 2.0 mm, TightSpec
matrix   80/100 Carbopack C support
matrix active group   0.1% SP-1000 phase
column type   packed GC

Description

General description

Use a glass column for applications where high inertness is required, such as for the analysis of active analytes. All glass columns undergo a proprietary high temperature silanization to ensure inertness. This column fits Agilent 5880, 5890, 6890. 7820, and 7890 (configuration A, on-column injection, all detectors except TCD).

Other Notes

This is a TightSpec column, which conforms to within +/- 6 mm of its stated length.

Price and Availability

Kit component also available separately

Description

Product #

Add to Cart

Empty Glass GC Column, for Agilent® 5880, 5890, 6890, 7820, and 7890 (Configuration "A"), L × O.D. × I.D. 6.6 ft (2.0 m) × 1/4 in. × 2.0 mm, TightSpec SDS 21814
Coated GC Packing, phase 0.1% SP-1000, matrix 80/100 Carbopack C support, bottle of 15 g SDS 11820
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products
 in. × 2.0 mm, TightSpec#@#Coated GC Packing, phase 0.1% SP-1000, matrix 80/100 Carbopack C support, bottle of 15 g" />

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?