• USA Home
  • 28513-U - SLB®-5ms Capillary GC Column

EMAIL THIS PAGE TO A FRIEND
28513-U Supelco

SLB®-5ms Capillary GC Column

L × I.D. 30 m × 0.20 mm, df 0.20 μm

  •  eCl@ss 32119290

Purchase

Properties

Related Categories β-Glucuronidase Drug Analysis Bundle, Analytical and Industrial Enzymes, Analytical/Chromatography, Biochemicals and Reagents, Capillary GC Columns and Guard Columns/Retention Gaps,
material   fused silica
Beta value   250
df   0.20 μm
application(s)   gas chromatography (GC): suitable
L × I.D.   30 m × 0.20 mm
matrix active group   Bonded and highly crosslinked; silphenylene polymer virtually equivalent in polarity to poly(5% diphenyl/95% dimethyl siloxane) phase
column type   capillary non-polar

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

General description

Application: The 5% phenyl equivalent phase provides a boiling point elution order with a slight increase in selectivity, especially for aromatic compounds. The low bleed characteristics, inertness, and durable nature make it the column of choice for environmental analytes (such as semivolatiles, pesticides, PCBs, and herbicides) or anywhere a low bleed non-polar column is required.
USP Code: This column meets USP G27 and G36 requirements.
Phase:
• Bonded and highly crosslinked
• Silphenylene polymer virtually equivalent in polarity to poly(5% diphenyl/95% dimethyl siloxane)
Temp. Limits:
• ≤0.32 mm I.D.: -60 °C to 340 °C (isothermal) or 360 °C (programmed)
• ≥0.53 mm I.D.: -60 °C to 330 °C (isothermal) or 340 °C (programmed)

Legal Information

SLB is a registered trademark of Sigma-Aldrich Co. LLC

Other Notes

Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Articles

Melamine and Related Compounds in Dog Food Using GC-MS

Melamine contamination in food became an issue in recent years after the discovery of it, and related compounds, in pet food and baby formula. It was discovered that melamine was intentionally added ...
Katherine K. Stenerson, Michael D. Buchanan
Reporter US Volume 28.3
Keywords: Centrifugation, Chromatography, Gas chromatography, Gas chromatography mass spectrometry, High performance liquid chromatography, Mass spectrometry, Positron Emission Tomography, Sonication

Related Content

How to Choose a Capillary GC Column

An optimized chromatographic separation begins with the column. The selection of the proper capillary column for any application should be based on four significant factors: stationary phase, column ...
How to Choose a Capillary GC Column
Keywords: Chromatography, Condensations, Gas chromatography, Immobilization, Mass spectrometry, Pesticides, Separation, Size-exclusion chromatography, Solid phase microextractions, Solvents, Vaporization

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?