-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
| Related Categories | β-Glucuronidase, β-Glucuronidase Drug Analysis Bundle, Analytical Enzymes, Analytical/Chromatography, Biochemicals and Reagents, |
| material | fused silica |
| Beta value | 250 |
| df | 0.32 μm |
| L × I.D. | 30 m × 0.32 mm |
| matrix active group | Bonded and highly crosslinked; silphenylene polymer virtually equivalent in polarity to poly(5% diphenyl/95% dimethyl siloxane) phase |
Live Chat and Frequently Asked Questions
are available for this Product.
Application: The 5% phenyl equivalent phase provides a boiling point elution order with a slight increase in selectivity, especially for aromatic compounds. The low bleed characteristics, inertness, and durable nature make it the column of choice for environmental analytes (such as semivolatiles, pesticides, PCBs, and herbicides) or anywhere a low bleed non-polar column is required.
USP Code: This column meets USP G27 and G36 requirements.
Phase:
• Bonded and highly crosslinked
• Silphenylene polymer virtually equivalent in polarity to poly(5% diphenyl/95% dimethyl siloxane)
Temp. Limits:
• ≤0.32 mm I.D.: -60 °C to 340 °C (isothermal) or 360 °C (programmed)
• ≥0.53 mm I.D.: -60 °C to 330 °C (isothermal) or 340 °C (programmed)
SLB is a trademark of Sigma-Aldrich Co. LLC
Take advantage of the Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?