• USA Home
  • 29026-U - Inter-Seal bonded caps (inseparable caps and liner), pkg of 100
EMAIL THIS PAGE TO A FRIEND
29026-U Supelco
Inter-Seal bonded caps (inseparable caps and liner), pkg of 100
black cap, PTFE/silicone septum, thread 9 mm
Purchase
Properties
Related Categories Alliance, Analytical/Chromatography, Autosampler/Vial Cross Reference, Caps, Crimp Seals and Stoppers, Inter-Seal Bonded Caps,
material   PTFE/silicone septum
  black cap
packaging   pkg of 100 ea
fitting   thread 9 mm
Description
General description
Inter-Seal bonded caps eliminate expensive and time consuming hand assembly while preventing liner fallout. They are made by a bonding process that bonds PTFE/silicone into polypropylene closures without using adhesives. The bonding properly seats and secures the septa within the vial opening. This integrated one piece system withstands multiple injections, making it very cost effective.
Benefits include:
•  Significant bond strength
•  Withstands multiple injections
•  Resistant to coring
•  Excellent resealability
•  Broad chemical resistance
Legal Information
Inter-Seal is a trademark of Integrated Liner Technologies, Inc.
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
29004-U Vials, screw top, R.A.M. (9 mm thread), large opening, 12 x 32 mm, volume 2 mL, amber glass vial, pkg of × 100 ea
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?