• USA Home
  • 29189-U - TFE Funnel for screw top vials
EMAIL THIS PAGE TO A FRIEND
29189-U Supelco
TFE Funnel for screw top vials
Kit with eight sizes
Purchase
Properties
Related Categories Analytical/Chromatography, TFE Funnels, Vial Accessories, Vials
material   white TFE
packaging   1 ea of  kit
compatibility   for use with screw thread vials (8/425 to 24/400 thread)
Description
General description
Our new threaded TFE funnels eliminate the mess that results from poor filing technique. The funnels are threaded on the inside of the base to allow them to screw onto the thread of the vial. The funnel then guides the syringe into the center of the vial where the sample is safely deposited, reducing the possibility of the sample running down the outside of the vial wall. The TFE funnel′s inert composition ensures that no sample contamination will occur.
TFE funnels are available to fit most standard vial sizes ranging from 2 mL to 60 mL vials. They may be ordered individually or as part of a variety kit.
TFE Vial Funnel Kit connects to threaded screw cap vials. The polypropylene kit contains one each of eight funnel sizes. The sizes included in this kit include:
• 8-425 2 mL standard opening vials
• 9-425 or 10/425 2 mL large opening vials
• 13-425 4 mL vial
• 15-425 7 mL vial
• 18-400 15 mL vial
• 20-400 22 mL vial
• 22-400 SPME vial
• 24-400. 20,40, or 60 mL vial
Other Notes
Vial not included.
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
29191-U TFE Funnel for screw top vials, for use with 2mL standard opening vial, 8-425 thread, pkg of 1 ea
29192-U TFE Funnel for screw top vials, for use with 9-425 and 10-425 threaded (2 mL large opening vials), pkg of 1 ea
29193-U TFE Funnel for screw top vials, for use with 13-425 thread, pkg of 1 ea
29194-U TFE Funnel for screw top vials, for use with 15-425 threaded vial, pkg of 1 ea
29196-U TFE Funnel for screw top vials, for use with 18-400 Thread (15 mL vial), pkg of 1 ea
29197-U TFE Funnel for screw top vials, for use with 22 mL threaded vials (20-400 thread), pkg of 1 ea
29198-U TFE Funnel for screw top vials, for use with SPME vial with 22-400 thread, pkg of 1 ea
29199-U TFE Funnel for screw top vials, for use with 20, 40, and 60 mL vials (24-400 thread), pkg of 1 ea
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?