• USA Home
  • 29194-U - TFE Funnel for screw top vials
EMAIL THIS PAGE TO A FRIEND
29194-U Supelco
TFE Funnel for screw top vials
for use with 15-425 threaded vial, pkg of 1 ea
Purchase
Properties
Related Categories Analytical/Chromatography, TFE Funnels, Vial Accessories, Vials
material   white TFE
packaging   pkg of 1 ea
opening size   1.4 in.
fitting   thread 15-425
compatibility   for use with 15-425 threaded vial
Description
General description
Our new threaded TFE funnels eliminate the mess that results from poor filing technique. The funnels are threaded on the inside of the base to allow them to screw onto the thread of the vial. The funnel then guides the syringe into the center of the vial where the sample is safely deposited, reducing the possibility of the sample running down the outside of the vial wall. The TFE funnel′s inert composition ensures that no sample contamination will occur.
TFE funnels are available to fit most standard vial sizes ranging from 2 mL to 60 mL vials. They may be ordered individually or as part of a variety kit.
Other Notes
Vial not included.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
27025 Vials, clear glass, screw top, graduated (vial only), volume 7 mL, thread 15-425, pkg of 100 ea
27072-U Vials, screw top, amber glass (vial only), volume 7 mL, amber glass vial, thread 15-425, pkg of 100 ea
27151 Vials, screw top, clear glass (vial only), volume 7 mL, clear glass vial, thread 15-425, pkg of 100 ea
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?