• USA Home
  • 29579-U - CGA-350 Gas Cylinder Pressure Regulator

EMAIL THIS PAGE TO A FRIEND
29579-U Supelco

CGA-350 Gas Cylinder Pressure Regulator

1-stage high purity plus, parameter 0-100 psi

Purchase

Description

General description

For use with hydrogen, methane, or 5% methane in argon gas cylinders.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?