• USA Home
  • 29615-U - SGE Autosampler Syringe, Gas Tight, Removable Needle, Dual Gauge
EMAIL THIS PAGE TO A FRIEND
29615-U Supelco
SGE Autosampler Syringe, Gas Tight, Removable Needle, Dual Gauge
volume 10 μL, 10R-HP-GT-63/47, needle size 23-26 ga (cone tip), needle L 42 mm
Purchase
Properties
Related Categories Agilent 7673/7683, Analytical/Chromatography, GC Autosampler Syringes, SGE Color-Coded Syringes for Agilent / HP 7673 & 7683 Autosampler, SGE MicroVolume syringes for autosamplers,
material   stainless steel needle cone tip
needle size   23-26 ga (cone tip)
description   color (dark orange)
feature   needle type RN (removable needle)
  point style 1
packaging   pkg of 1 ea
mfr. no.   SGE, 002829
needle L   42 mm
volume   10 μL
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?