• USA Home
  • 29619-U - Syringes for Shimadzu AOC14, 17, 20 and 20i Autosamplers
EMAIL THIS PAGE TO A FRIEND
29619-U Supelco
Syringes for Shimadzu AOC14, 17, 20 and 20i Autosamplers
5F-S-0.47, volume 5 μL, needle size 26 ga (cone tip), needle L 42 mm, color (lime)
Purchase
Properties
Related Categories Analytical/Chromatography, GC Autosampler Syringes, Shimadzu AOC14, AOC17, AOC20 GC Autosamplers, Syringes
material   stainless steel needle cone tip
needle size   26 ga (cone tip)
feature   needle type F (cemented needle)
  point style 1
packaging   pkg of 1 ea
mfr. no.   SGE, 001987
needle L   42 mm
volume   5 μL
Description
Legal Information
Shimadzu is a trademark of Shimadzu Corporation
Price and Availability
Safety & Documentation
Safety Information
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?