• USA Home
  • 29636-U - SGE autosampler syringes, removable needle, for CTC Analytics
EMAIL THIS PAGE TO A FRIEND
29636-U Supelco
SGE autosampler syringes, removable needle, for CTC Analytics
10R-C/T-GT-5/0.47C, Slim line, Removable needle, volume 10 μL, needle size 26 ga (cone tip), needle L × O.D. 50 mm × 0.47 mm
Purchase
Properties
Related Categories Analytical/Chromatography, CTC/Leap Technologies Autosamplers, GC Autosampler Syringes, SGE Color-Coated Autosampler syringes, SGE GC Syringes for CTC Analytics, Removable Needle,
material   stainless steel needle cone tip
needle size   26 ga (cone tip)
description   color (dark orange)
feature   syringe type R (removable needle)
packaging   pkg of 1 ea
mfr. no.   SGE, 002985
needle L × O.D.   50 mm × 0.47 mm
volume   10 μL
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?