• USA Home
  • 2X8200 - Dowex® 2X8 chloride form

EMAIL THIS PAGE TO A FRIEND
2X8200 Supelco

Dowex® 2X8 chloride form

100-200 mesh

Purchase

Properties

composition   Moisture content, 34-40%
cross-linkage   8%
matrix   styrene-divinylbenzene (gel)
matrix active group   dimethylethanolbenzyl ammonium functional group
particle size   100-200 mesh
capacity   1.2 meq/mL total capacity

Description

Application

Strongly basic (type II) anion exchange resin that meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Other Notes

This resin has not been specially prepared for use. We suggest that it be subjected to a suitable preliminary elution and wash.

Legal Information

DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?