• USA Home
  • 31814 - Molecular sieves, 5 Å

EMAIL THIS PAGE TO A FRIEND
31814 Supelco

Molecular sieves, 5 Å

for GC, 0.5 nm

Synonym: Calcium, Sodium Alumino-Silicate

  • CAS Number 69912-79-4

  • Linear Formula CanNa12-2n[(AlO2)12(SiO2)12] · xH2O

  •  MDL number MFCD00147627

  •  eCl@ss 32110204

Purchase

Properties

grade   for GC
quality   0.5 nm
granulation   ~0.5 mm

Description

Application

Molecules with an effective diameter of 5Å, including C3H8 n-C4H10 to C22H46 and n-C4H9 are adsorbed.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?