• USA Home
  • 33045-U - Methoxyamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
33045-U Supelco

Methoxyamine hydrochloride

bottle of 5 g

Synonym: O-Methylhydroxylamine hydrochloride, Methoxylamine hydrochloride, O-Methoxyamine HCl

Purchase

Properties

Related Categories Alkylation Reagents, Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, General Alkylation Reagents More...
description   Alkylation reagent
InChI Key   XNXVOSBNFZWHBV-UHFFFAOYSA-N
packaging   bottle of 5 g
mp   151-154 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN3261 - class 8 - PG 2 - EHS - Corrosive solid, acidic, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
NC3980000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?