• USA Home
  • 33806 - Sulphated β-cyclodextrin, 100mg, neat

EMAIL THIS PAGE TO A FRIEND
33806 Supelco

Sulphated β-cyclodextrin, 100mg, neat

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Chiral, Derivatization Reagents, Derivatization Reagents HPLC More...
packaging   box of 1 ampule
concentration   100 mg/ampule neat

Description

Application

It was used as a chiral selector in enantioselective analysis during disopyramide and mono-N-dealkyldisopyramide enantiomer determination.

General description

Sulphated β-cyclodextrin is an anionic derivative class of cyclodextrin. It has been used in capillary electrophoresis. It was considered as chiral stationary phase to enantioresolve. Also used as chiral mobile phase additive. It is shown to have mobility opposite to the electrosmotic flow and shows strong resolving power at low concentrations.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?