• USA Home
  • 33807-U - Carboxylated β-cyclodextrin, 100mg, neat

EMAIL THIS PAGE TO A FRIEND
33807-U Supelco

Carboxylated β-cyclodextrin, 100mg, neat

Purchase

Properties

packaging   box of 1 ampule
concentration   100 mg/ampule neat
storage temp.   room temp

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?