EMAIL THIS PAGE TO A FRIEND
40087 Supelco

Endrin solution

certified reference material, 5000 μg/mL in methanol

Purchase

Description

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
2
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Multiple Pesticides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Multiple Pesticides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Keywords: Agriculture, Chromatography, Gas chromatography, Metabolites, Pesticides, Purification, Solid phase microextractions

GC Analysis of Organochlorine Pesticides in Water on SPB®-5 after SPME using 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Organochlorine Pesticides in Water on SPB®-5 after SPME using 100 μm PDMS Fiber
Keywords: Chromatography, Electron capture detector, Environmental, Gas chromatography, Metabolites, Pesticides, Purification, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of Semivolatiles in Drinking Water on a 5% Phenyl Column after SPE using ENVI™-18 DSK

From our library of Articles, Sigma-Aldrich presents GC Analysis of Semivolatiles in Drinking Water on a 5% Phenyl Column after SPE using ENVI™-18 DSK
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Sample preparations, Solid phase extractions

GC Analysis of a 20-Component Organochlorine Pesticide Mix in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI™-8

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 20-Component Organochlorine Pesticide Mix in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI™-8
Keywords: Chromatography, Electron capture detector, Gas chromatography, Purification, Sample preparations, Size-exclusion chromatography, Solid phase extractions

GC Analysis of a 7-Component Organochlorine Pesticide Mix in Drinking Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI™-18

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 7-Component Organochlorine Pesticide Mix in Drinking Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI™-18
Keywords: Chromatography, Electron capture detector, Gas chromatography, Metabolites, Pesticides, Purification, Sample preparations, Solid phase extractions

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on Equity®-1701 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on Equity®-1701 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SPB®-608

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SPB®-608
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification

US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8081: GC Analysis of Organochlorine Pesticides on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8081: GC Analysis of Organochlorine Pesticides on Equity®-5
Keywords: Chromatography, Electron capture detector, Environmental, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?