• USA Home
  • 40177 - 1,2,4,5-Tetrachlorobenzene solution

EMAIL THIS PAGE TO A FRIEND
40177 Supelco

1,2,4,5-Tetrachlorobenzene solution

certified reference material, 1000 μg/mL in acetonitrile

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   certified reference material
  TraceCERT®
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   1000 μg/mL in acetonitrile
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?