• USA Home
  • 40349 - 4-Methyl-2-pentanone solution

EMAIL THIS PAGE TO A FRIEND
40349 Supelco

4-Methyl-2-pentanone solution

certified reference material, 5000 μg/mL in methanol

Synonym: Methyl isobutyl ketone

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Certified Reference Materials (CRMs),
grade   certified reference material
  TraceCERT®
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   5000 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Class 3 Residual Solvents on Equity®-1 after SPME using 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Class 3 Residual Solvents on Equity®-1 after SPME using 100 μm PDMS Fiber
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pharmaceutical, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of Class 3 Residual Solvents on Equity®-5

From our library of Articles, Sigma-Aldrich presents GC Analysis of Class 3 Residual Solvents on Equity®-5
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pharmaceutical, Size-exclusion chromatography

GC Analysis of Class 3 Residual Solvents on OVI-G43

From our library of Articles, Sigma-Aldrich presents GC Analysis of Class 3 Residual Solvents on OVI-G43
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pharmaceutical, Size-exclusion chromatography

GC Analysis of Class 3 Residual Solvents on SUPELCOWAX® 10

From our library of Articles, Sigma-Aldrich presents GC Analysis of Class 3 Residual Solvents on SUPELCOWAX® 10
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pharmaceutical, Size-exclusion chromatography

GC Analysis of Class 3 Residual Solvents on VOCOL® after SPME using 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Class 3 Residual Solvents on VOCOL® after SPME using 100 μm PDMS Fiber
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pharmaceutical, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of Industrial Solvents on Equity®-1701

From our library of Articles, Sigma-Aldrich presents GC Analysis of Industrial Solvents on Equity®-1701
Keywords: Chromatography, Flame ionization detector, Gas chromatography

GC Analysis of Industrial Solvents on Equity®-1

From our library of Articles, Sigma-Aldrich presents GC Analysis of Industrial Solvents on Equity®-1
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Industrial Solvents on SUPELCOWAX 10

From our library of Articles, Sigma-Aldrich presents GC Analysis of Industrial Solvents on SUPELCOWAX 10
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Residual Solvents on Equity®-5

From our library of Articles, Sigma-Aldrich presents GC Analysis of Residual Solvents on Equity®-5
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pharmaceutical, Purification, Size-exclusion chromatography

GC Analysis of Volatiles in Milk (Before and After Exposure to Sunlight) on Supel-Q™ PLOT after SPME using a 75 μm PDMS/Carboxen Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Volatiles in Milk (Before and After Exposure to Sunlight) on Supel-Q™ PLOT after SPME using a 75 μm PDMS/Carboxen Fiber
Keywords: Chromatography, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Purification, Sample preparations, Separation, Solid phase microextractions

GC Analysis of a 56-Component Industrial Solvents Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 56-Component Industrial Solvents Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column
Keywords: Chromatography, Flame ionization detector, Gas chromatography

GC Analysis of an Indoor Air Standard on Equity®-1

From our library of Articles, Sigma-Aldrich presents GC Analysis of an Indoor Air Standard on Equity®-1
Keywords: Air Monitoring, Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8015: GC Analysis of Nonhalogenated Volatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8015: GC Analysis of Nonhalogenated Volatiles on SLB®-5ms
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

US EPA Method 8260: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8260: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Purge and trap, Purification, Solid phase microextractions

US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification, Solid phase microextractions

US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap
Keywords: Chromatography, Gas chromatography, Purification, Solid phase microextractions

US EPA Method OLM04.2 VOA: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 VOA: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis
Keywords: Chromatography, Environmental, Gas chromatography, Purification, Solid phase microextractions

US EPA Method OLM04.2 VOA: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 VOA: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis
Keywords: Chromatography, Environmental, Gas chromatography, Purification, Solid phase microextractions

US EPA Method TO-17: GC Analysis of Volatiles on Equity®-1 after Collection/Desorption using Carbotrap 300 Tube

From our library of Articles, Sigma-Aldrich presents US EPA Method TO-17: GC Analysis of Volatiles on Equity®-1 after Collection/Desorption using Carbotrap 300 Tube
Keywords: Chromatography, Gas chromatography

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?