-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
CAS Number 59-89-2
EC Number 200-659-6
| Related Categories | Alpha Sort, Alphabetic, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | analytical standard |
| packaging | ampule of 1 mL |
| concentration | 5000 μg/mL in methanol |
| storage temp. | 2-8°C |
Customers Also Purchased
5000 μg/mL in methanol, analytical standard
5000 μg/mL in methanol, analytical standard
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?