• USA Home
  • 41112 - Propylbenzene solution

EMAIL THIS PAGE TO A FRIEND
41112 Supelco

Propylbenzene solution

certified reference material, 5000 μg/mL in methanol

Synonym: 1-Phenylpropane

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   certified reference material
  TraceCERT®
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   5000 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Hydrocarbons in Gasoline on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Hydrocarbons in Gasoline on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Size-exclusion chromatography

GC Analysis of Refinery Grade Unleaded Gasoline on TCEP (15 m x 0.10 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Refinery Grade Unleaded Gasoline on TCEP (15 m x 0.10 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Volatiles on SPB®-624, Direct Injection

From our library of Articles, Sigma-Aldrich presents GC Analysis of Volatiles on SPB®-624, Direct Injection
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Volatiles on VOCOL® after SPME using 75 μm Carboxen/PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Volatiles on VOCOL® after SPME using 75 μm Carboxen/PDMS Fiber
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Solid phase microextractions

US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, ELCD Signal

From our library of Articles, Sigma-Aldrich presents US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, ELCD Signal
Keywords: Chromatography, Gas chromatography, Hall electrolytic conductivity detector, Photo-ionization detector, Purge and trap, Sample preparations

US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, PID Signal

From our library of Articles, Sigma-Aldrich presents US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, PID Signal
Keywords: Chromatography, Gas chromatography, Hall electrolytic conductivity detector, Photo-ionization detector, Purification

US EPA Method 524.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 524.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification, Solid phase microextractions

US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap
Keywords: Chromatography, Gas chromatography, Purification, Solid phase microextractions

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?