EMAIL THIS PAGE TO A FRIEND
442244 Supelco

1,3-Dinitrobenzene

analytical standard

  • CAS Number 99-65-0

  • Linear Formula C6H4(NO2)2

  • Molecular Weight 168.11

  •  Beilstein Registry Number 1105654

  •  EC Number 202-776-8

  •  MDL number MFCD00007222

  •  PubChem Substance ID 24867752

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Aromatics, Chemical Class,
grade   analytical standard
InChI Key   WDCYWAQPCXBPJA-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   297 °C(lit.)
mp   84-86 °C(lit.)
density   1.368 g/mL at 25 °C(lit.)
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3443 6.1 / PGII
WGK Germany 
3
RTECS 
CZ7350000
Flash Point(F) 
302 °F
Flash Point(C) 
150 °C
Protocols & Articles

Articles

EPA Method 8330: HPLC Analysis of Explosives on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents EPA Method 8330: HPLC Analysis of Explosives on Ascentis® Express C18
Keywords: Chromatography, High performance liquid chromatography

GC Analysis of Explosives in Water on a 14% Cyanopropylphenyl Column after SPME using 65 μm PDMS/DVB Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Explosives in Water on a 14% Cyanopropylphenyl Column after SPME using 65 μm PDMS/DVB Fiber
Keywords: Chromatography, Electron capture detector, Gas chromatography, Purification, Solid phase microextractions

LC-MS Analysis of Explosives on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents LC-MS Analysis of Explosives on Ascentis® Express C18
Keywords: Chromatography, High performance liquid chromatography, Mass spectrometry

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8330: HPLC Analysis of Nitroaromatics and Nitramines, Mix A on Ascentis® C18

From our library of Articles, Sigma-Aldrich presents US EPA Method 8330: HPLC Analysis of Nitroaromatics and Nitramines, Mix A on Ascentis® C18
Keywords: Chromatography, Environmental, Environmental Analysis, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?