Synonym: Tetramethylene dichloride
CAS Number 110-56-5
Linear Formula Cl(CH2)4Cl
Molecular Weight 127.01
Beilstein Registry Number 635673
EC Number 203-778-1
MDL number MFCD00001011
PubChem Substance ID 24867754
| Related Categories | Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography, |
| grade | analytical standard |
| packaging | ampule of 1000 mg |
| refractive index |
n |
| bp | 161-163 °C(lit.) |
| mp | −38 °C(lit.) |
| density | 1.16 g/mL at 25 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?