EMAIL THIS PAGE TO A FRIEND
442350 Supelco

2-Fluorophenol

analytical standard

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chemical Class,
grade   analytical standard
InChI Key   HFHFGHLXUCOHLN-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.511(lit.)
bp   171-172 °C/741 mmHg(lit.)
mp   16.1 °C(lit.)
density   1.256 g/mL at 25 °C(lit.)
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
RTECS 
SL4530000
Flash Point(F) 
116.6 °F
Flash Point(C) 
47 °C
Protocols & Articles

Articles

Phenolic Calibration Standards

Phenolic compounds are important raw materials for the manufacturing of phenolic resins, pharmaceuticals, laboratory reagents, dyes, germicidal paints, and general disinfectants. Due to their toxicit...
Reporter US Volume 28.3
Keywords: Chromatography, Environmental, Gas chromatography, Pharmaceutical, Sample preparations

US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.
Keywords: Chromatography, Gas chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?