EMAIL THIS PAGE TO A FRIEND
442362 Supelco

2-Naphthylamine

analytical standard

Synonym: β-Naphthylamine, 2-Aminonaphthalene

  • CAS Number 91-59-8

  • Linear Formula C10H7NH2

  • Molecular Weight 143.19

  •  Beilstein Registry Number 3939429

  •  EC Number 202-080-4

  •  MDL number MFCD00004112

  •  PubChem Substance ID 24867814

Purchase

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1650 6.1 / PGII
WGK Germany 
3
RTECS 
QM2100000
Protocols & Articles

Articles

GC Analysis of Semivolatiles on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Semivolatiles on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Keywords: Chromatography, Gas chromatography, Purification, Solid phase microextractions

GC Analysis of a 32-Component Anilines Mix on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 32-Component Anilines Mix on SLB®-IL59
Keywords: Chromatography, Gas chromatography, Mass selective detector

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?