EMAIL THIS PAGE TO A FRIEND
442375 Supelco

3,4-Dichlorophenol

analytical standard

  • CAS Number 95-77-2

  • Linear Formula Cl2C6H3OH

  • Molecular Weight 163.00

  •  Beilstein Registry Number 1907693

  •  EC Number 202-450-5

  •  MDL number MFCD00002258

  •  PubChem Substance ID 24867819

Purchase

Properties

Related Categories A-D, Additional Standards, All Herbicide Standards, Alpha Sort, Analytical Standards,
grade   analytical standard
InChI Key   WDNBURPWRNALGP-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   145-146 °C(lit.)
mp   65-67 °C(lit.)
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2020 6.1 / PGIII
WGK Germany 
3
Protocols & Articles

Articles

Phenolic Calibration Standards

Phenolic compounds are important raw materials for the manufacturing of phenolic resins, pharmaceuticals, laboratory reagents, dyes, germicidal paints, and general disinfectants. Due to their toxicit...
Reporter US Volume 28.3
Keywords: Chromatography, Environmental, Gas chromatography, Pharmaceutical, Sample preparations

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?